Assay Details
| Gene Symbol: Slc7a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011990.2 | NP_036120.1 | 10 - 11 | 1622 | 73 | |
| GenBank mRNA | AB022345.1 | - | 10 - 11 | 1589 | 73 | |
| AK076322.1 | - | 10 - 11 | 1597 | 73 | ||
| AK089202.1 | - | 10 - 11 | 1595 | 73 | ||
| AK162233.1 | - | 10 - 11 | 1397 | 73 | ||
| AK163168.1 | - | 10 - 11 | 1596 | 73 | ||
| AK165599.1 | - | 10 - 11 | 1598 | 73 | ||
| AK171192.1 | - | 10 - 11 | 1620 | 73 | ||
| AK171252.1 | - | 10 - 11 | 1596 | 73 | ||
| AY766236.1 | - | 10 - 11 | 1589 | 73 | ||
| AY766237.1 | - | 1589 | 73 | |||
| BC141402.1 | - | 10 - 11 | 1519 | 9056015 | 73 | |
| BC141408.1 | - | 10 - 11 | 1519 | 9056021 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a11
Genomic Map