Assay Details
| Gene Symbol: Spg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144987.1 | NP_001138459.1 | 4 - 5 | 1513 | 82 | |
| NM_144895.2 | NP_659144.1 | 4 - 5 | 1297 | 82 | ||
| GenBank mRNA | AK031545.1 | - | 3 - 4 | 852 | 82 | |
| AK087388.1 | - | 4 - 5 | 1266 | 82 | ||
| AK089946.1 | - | 4 - 5 | 1298 | 82 | ||
| AK147760.1 | - | 4 - 5 | 1515 | 82 | ||
| AK159951.1 | - | 4 - 5 | 1298 | 82 | ||
| AK192376.1 | - | 2 - 3 | 199 | 82 | ||
| BC022921.1 | - | 4 - 5 | 1265 | 4220506 | 82 | |
More Information
Set Membership: |
|

Spg20
Genomic Map