Assay Details
| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013529.3 | NP_038557.1 | 4 - 5 | 417 | 118 | |
| GenBank mRNA | AB016780.1 | - | 4 - 5 | 383 | 118 | |
| AK132821.1 | - | 3 - 4 | 263 | 118 | ||
| BC031928.1 | - | 4 - 5 | 385 | 4167189 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfpt2
Genomic Map