Assay Details
| Gene Symbol: Ttc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133795.1 | NP_598556.1 | 6 - 7 | 766 | 171 | |
| GenBank mRNA | AK077715.1 | - | 6 - 7 | 779 | 171 | |
| AK166431.1 | - | 6 - 7 | 773 | 171 | ||
| AK170230.1 | - | 6 - 7 | 776 | 171 | ||
| BC010236.1 | - | 6 - 7 | 766 | 3581904 | 171 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc1
Genomic Map