Assay Details
| Gene Symbol: Adra1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284381.1 | NP_001271310.1 | 2 - 3 | 297 | 95 | |
| GenBank mRNA | AK004891.1 | - | 2 - 3 | 254 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004891.1, NM_001284381.1 transcripts.
Set Membership: |
|

Adra1b
Genomic Map