Assay Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008485.3 | NP_032511.3 | 6 - 7 | 943 | 66 | |
| GenBank mRNA | L20477.1 | - | 4 - 5 | 503 | 66 | |
| U43327.2 | - | 5 - 6 | 676 | 66 | ||
More Information
Set Membership: |
|

Lamc2
Genomic Map