Assay Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008485.3 | NP_032511.3 | 9 - 10 | 1373 | 68 | |
| GenBank mRNA | L20477.1 | - | 7 - 8 | 933 | 68 | |
| U43327.2 | - | 8 - 9 | 1106 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lamc2
Genomic Map