Assay Details
| Gene Symbol: Cdh20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011800.4 | NP_035930.1 | 10 - 11 | 1860 | 114 | |
| GenBank mRNA | AF007116.1 | - | 10 - 11 | 2150 | 114 | |
| AK034475.1 | - | 10 - 11 | 1861 | 114 | ||
| BC119606.2 | - | 10 - 11 | 1839 | 40061260 | 114 | |
| BC119607.2 | - | 10 - 11 | 1839 | 40061261 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh20
Genomic Map