Assay Details
| Gene Symbol: Cdh20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011800.4 | NP_035930.1 | 5 - 6 | 1044 | 91 | |
| GenBank mRNA | AF007116.1 | - | 5 - 6 | 1334 | 91 | |
| AK034475.1 | - | 5 - 6 | 1045 | 91 | ||
| BC119606.2 | - | 5 - 6 | 1023 | 40061260 | 91 | |
| BC119607.2 | - | 5 - 6 | 1023 | 40061261 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh20
Genomic Map