Assay Details
| Gene Symbol: Slc7a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172861.3 | NP_766449.1 | 6 - 7 | 1414 | 91 | |
| GenBank mRNA | AK044448.1 | - | 6 - 7 | 1407 | 91 | |
| AK163358.1 | - | 6 - 7 | 1458 | 91 | ||
| AK173217.1 | - | 6 - 7 | 1390 | 91 | ||
| BC061928.1 | - | 6 - 7 | 1390 | 5359765 | 91 | |
More Information
Set Membership: |
|

Slc7a14
Genomic Map