Assay Details
| Gene Symbol: Slamf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029612.4 | NP_083888.3 | 3 - 4 | 790 | 124 | |
| GenBank mRNA | AB196824.1 | - | 3 - 4 | 653 | 124 | |
| AB196825.1 | - | 3 - 4 | 653 | 124 | ||
| AB196826.1 | - | 3 - 4 | 653 | 124 | ||
| AB196827.1 | - | 3 - 4 | 653 | 124 | ||
| AK009505.1 | - | 3 - 4 | 746 | 124 | ||
| AK090041.1 | - | 3 - 4 | 781 | 124 | ||
| BC019477.1 | - | 3 - 4 | 729 | 4458767 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | BC019477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | BE447476.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | AK009505.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | AK090041.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | NM_029612.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | BC019477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | BE447476.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | AK009505.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | AK090041.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246066 | NM_029612.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97876 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97222 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162093 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | BC019477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | BE447476.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | AK009505.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | AK090041.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | NM_029612.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | BC019477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | BE447476.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | AK009505.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | AK090041.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246066 | NM_029612.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20833215 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162093 | - | - |
| Whole Mouse Genome G4122A | A_51_P246066 | BC019477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | BE447476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | AK009505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | AK090041.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | NM_029612.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | BC019477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | BE447476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | AK009505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | AK090041.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246066 | NM_029612.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116574 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162093 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00290568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00290550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00290565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00290558 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00290560 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00290559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20833270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00550938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00742678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00742686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00290560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00733302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00225602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01134401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00131730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01134410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00290550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00290566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01134405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01134403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00290565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20833248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351794 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419315_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351792 | - | - |
| Mouse Genome 430 2.0 Array | 1419315_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419315_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98365_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98365_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100013427.mm.1 | NM_029612.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013427.mm.1 | NM_029612.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013427.mm.1 | TC0100001615.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419315_at | - | - |
| Mouse Expression Set 430 | 1419315_PM_at | - | - |
| Murine Genome U74v2 Set | 169975_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169975_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419315_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419315_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419315_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419315_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3710300 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710300 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663249 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2663249 | - | - |
| mouse-6 v1.1 (Illumina) | scl17201.4.1_50-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710300 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17201.4.1_50-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2663249 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2663249 | - | - |

Slamf9
Genomic Map