Assay Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145800.1 | NP_001139272.1 | 2 - 3 | 316 | 62 | |
| NM_033608.3 | NP_291086.2 | 2 - 3 | 382 | 62 | ||
| GenBank mRNA | AF317839.1 | - | 2 - 3 | 382 | 62 | |
| AK053544.1 | - | 2 - 3 | 310 | 62 | ||
| AK078536.1 | - | 2 - 3 | 310 | 62 | ||
| AK145121.1 | - | 2 - 3 | 291 | 62 | ||
| BC034594.1 | - | 2 - 3 | 298 | 62 | ||
| BC040281.1 | - | 2 - 3 | 342 | 5364073 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf9
Genomic Map