Assay Details
| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028057.2 | NP_082333.1 | 7 - 8 | 692 | 81 | |
| GenBank mRNA | AK005159.1 | - | 7 - 8 | 693 | 81 | |
| AK159663.1 | - | 7 - 8 | 662 | 81 | ||
| AK167178.1 | - | 7 - 8 | 686 | 81 | ||
| BC016266.1 | - | 7 - 8 | 1488 | 4924006 | 81 | |
| BC024618.1 | - | 7 - 8 | 670 | 4235599 | 81 | |
| JX420288.1 | - | 772 | 81 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r1
Genomic Map