Assay Details
| Gene Symbol: Tmem183a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042485.1 | NP_001035950.1 | 5 - 6 | 825 | 88 | |
| NM_020588.2 | NP_065613.1 | 5 - 6 | 828 | 88 | ||
| GenBank mRNA | AB041592.1 | - | 5 - 6 | 784 | 88 | |
| AK004922.2 | - | 5 - 6 | 809 | 88 | ||
| AK007779.1 | - | 5 - 6 | 807 | 88 | ||
| AK152852.1 | - | 5 - 6 | 768 | 88 | ||
| AK168331.1 | - | 5 - 6 | 770 | 88 | ||
| BC003210.1 | - | 5 - 6 | 753 | 3156957 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem183a
Genomic Map