Assay Details
| Gene Symbol: Rhoj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023275.2 | NP_075764.1 | 3 - 4 | 908 | 87 | |
| GenBank mRNA | AB060651.1 | - | 3 - 4 | 813 | 87 | |
| AF309564.1 | - | 3 - 4 | 767 | 87 | ||
| AJ276568.1 | - | 3 - 4 | 367 | 87 | ||
| AK003482.1 | - | 3 - 4 | 796 | 87 | ||
| AK003490.1 | - | 3 - 4 | 796 | 87 | ||
| AK156619.1 | - | 3 - 4 | 796 | 87 | ||
| AK157518.1 | - | 3 - 4 | 796 | 87 | ||
| AK159385.1 | - | 3 - 4 | 829 | 87 | ||
| BC024626.1 | - | 3 - 4 | 788 | 87 | ||
| BC043719.1 | - | 3 - 4 | 845 | 5373358 | 87 | |
| BC054464.1 | - | 3 - 4 | 897 | 87 | ||
| BC060193.1 | - | 3 - 4 | 786 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rhoj
Genomic Map