Assay Details
| Gene Symbol: Rpl10l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162933.1 | NP_001156405.1 | 1 | 868 | 108 | |
| GenBank mRNA | BC052145.1 | - | 1 | 868 | 108 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl10l
Genomic Map