Assay Details
| Gene Symbol: Sertm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177854.4 | NP_808522.1 | 1 - 2 | 148 | 54 | |
| GenBank mRNA | AK031306.1 | - | 1 - 2 | 149 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031306.1, NM_177854.4 transcripts.
Set Membership: |
|

Sertm1
Genomic Map