Assay Details
| Gene Symbol: Pwwp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164231.1 | NP_001157703.1 | 1 - 2 | 623 | 152 | |
| NM_027557.1 | NP_081833.1 | 1 - 2 | 623 | 152 | ||
| GenBank mRNA | AK173305.1 | - | 1 - 2 | 615 | 152 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P108263 | BC044810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P108263 | AK171257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P108263 | NM_001164231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151363 | AK173305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151363 | AK050921.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151363 | NM_027557.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2151363 | AK014534.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156923 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P108263 | BC044810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P108263 | AK171257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P108263 | NM_001164231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151363 | AK173305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151363 | AK050921.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151363 | NM_027557.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2151363 | AK014534.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156923 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07322292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07322221 | - | - |
| Whole Mouse Genome G4122A | A_51_P215293 | AK173305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215293 | AK050921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215293 | NM_027557.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P215293 | AK014534.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09387424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09110760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09723778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09051669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09723783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09409938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09387449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27751828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08992652 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375328 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453028_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435633_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375326 | - | - |
| Mouse Genome 430 2.0 Array | 1435633_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453028_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453028_at | - | - |
| Mouse Genome 430 2.0 Array | 1435633_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70802_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70802_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100003366.mm.1 | NM_001164231.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003366.mm.1 | NM_001164231.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003366.mm.1 | NM_027557.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003366.mm.1 | NM_027557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003366.mm.1 | TC1100000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100003372.mm.1 | NM_027557.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003372.mm.1 | NM_027557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003372.mm.1 | TC1100000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100003370.mm.1 | NM_001164231.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003370.mm.1 | NM_001164231.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003370.mm.1 | NM_027557.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003370.mm.1 | NM_027557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003370.mm.1 | TC1100000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100003375.mm.1 | NM_001164231.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003375.mm.1 | NM_001164231.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003375.mm.1 | TC1100000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100003377.mm.1 | NM_001164231.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003377.mm.1 | NM_001164231.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003377.mm.1 | TC1100000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100003371.mm.1 | NM_001164231.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003371.mm.1 | NM_001164231.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003371.mm.1 | NM_027557.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003371.mm.1 | NM_027557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003371.mm.1 | TC1100000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100003364.mm.1 | NM_001164231.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003364.mm.1 | NM_001164231.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003364.mm.1 | NM_027557.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003364.mm.1 | NM_027557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003364.mm.1 | TC1100000432.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100003365.mm.1 | NM_001164231.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003365.mm.1 | NM_001164231.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003365.mm.1 | NM_027557.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003365.mm.1 | NM_027557.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003365.mm.1 | TC1100000432.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435633_at | - | - |
| Mouse Expression Set 430 | 1435633_PM_at | - | - |
| Mouse Expression Set 430 | 1453028_at | - | - |
| Mouse Expression Set 430 | 1453028_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453028_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435633_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435633_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453028_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 895936 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2427777 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256888 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2460900 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588040 | - | - |

Pwwp2a
Genomic Map