Assay Details
| Gene Symbol: Gpr33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008159.2 | NP_032185.1 | 1 - 2 | 126 | 67 | |
| GenBank mRNA | AF045766.1 | - | 1 - 2 | 149 | 67 | |
| BC064084.1 | - | 1 - 2 | 126 | 575313 | 67 | |
| GQ981323.1 | - | 1 - 2 | 167 | 67 | ||
| GQ981327.1 | - | 1 - 2 | 229 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF045766.1, BC064084.1, GQ981323.1, GQ981327.1, NM_008159.2 transcripts.
Set Membership: |
|

Gpr33
Genomic Map