Assay Details
| Gene Symbol: Fmod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021355.3 | NP_067330.1 | 1 - 2 | 60 | 60 | |
| GenBank mRNA | AK032302.1 | - | 1 - 2 | 61 | 60 | |
| AK053317.1 | - | 1 - 2 | 61 | 60 | ||
| AK054183.1 | - | 1 - 2 | 117 | 60 | ||
| AK080601.1 | - | 1 - 2 | 78 | 60 | ||
| AK082593.1 | - | 1 - 2 | 77 | 60 | ||
| BC064779.1 | - | 1 - 2 | 33 | 30058603 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032302.1, AK053317.1, AK054183.1, AK080601.1, AK082593.1, BC064779.1, NM_021355.3 transcripts.
Set Membership: |
|

Fmod
Genomic Map