Assay Details
| Gene Symbol: Fcrlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029984.2 | NP_001025155.2 | 4 - 5 | 570 | 63 | |
| GenBank mRNA | AY513660.1 | - | 4 - 5 | 570 | 63 | |
| DQ119292.1 | - | 4 - 5 | 570 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825280 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825266 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825271 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050872 | AY513660.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050872 | DQ119292.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050872 | NM_001029984.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139227 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050872 | AY513660.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050872 | DQ119292.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050872 | NM_001029984.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20825295 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04353067 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825280 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825266 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20825272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20825215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00534022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00533980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00351041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00775802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00846983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01141593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00534026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00816061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01055419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00835528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00572207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00906070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01055424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00854127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00709912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00572201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00572217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20825308 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360013 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360012 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100028788.mm.1 | NM_001029984.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028788.mm.1 | NM_001029984.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028788.mm.1 | TC0100003519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100028790.mm.1 | NM_001029984.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028790.mm.1 | NM_001029984.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028790.mm.1 | TC0100003519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100028787.mm.1 | NM_001029984.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028787.mm.1 | NM_001029984.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028787.mm.1 | TC0100003519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100028792.mm.1 | NM_001029984.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028792.mm.1 | NM_001029984.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028792.mm.1 | TC0100003519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100028789.mm.1 | NM_001029984.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028789.mm.1 | NM_001029984.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028789.mm.1 | TC0100003519.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 540150 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162188 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162188 | - | - |
| mouse-6 v1.1 (Illumina) | GI_71725378-S | - | - |

Fcrlb
Genomic Map