Assay Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008911.2 | NP_032937.1 | 3 - 4 | 543 | 95 | |
| GenBank mRNA | AK142632.1 | - | 3 - 4 | 495 | 95 | |
| BC002047.1 | - | 3 - 4 | 498 | 3484079 | 95 | |
| D45185.1 | - | 3 - 4 | 269 | 95 | ||
| U25114.1 | - | 3 - 4 | 518 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppox
Genomic Map