Assay Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008911.2 | NP_032937.1 | 8 - 9 | 1191 | 59 | |
| GenBank mRNA | AK134341.1 | - | 2623 | 59 | ||
| AK142632.1 | - | 8 - 9 | 1143 | 59 | ||
| BC002047.1 | - | 8 - 9 | 1146 | 3484079 | 59 | |
| D45185.1 | - | 8 - 9 | 917 | 59 | ||
| U25114.1 | - | 8 - 9 | 1166 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppox
Genomic Map