Assay Details
| Gene Symbol: Agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178767.5 | NP_848882.2 | 11 - 12 | 1566 | 73 | |
| GenBank mRNA | AK039191.1 | - | 11 - 12 | 1137 | 73 | |
| AK040702.1 | - | 11 - 12 | 1567 | 73 | ||
| AK163155.1 | - | 11 - 12 | 1347 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agmo
Genomic Map