Assay Details
| Gene Symbol: Agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178767.5 | NP_848882.2 | 4 - 5 | 915 | 122 | |
| GenBank mRNA | AK039191.1 | - | 4 - 5 | 486 | 122 | |
| AK040702.1 | - | 4 - 5 | 916 | 122 | ||
| AK050246.1 | - | 4 - 5 | 776 | 122 | ||
| AK163155.1 | - | 4 - 5 | 696 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agmo
Genomic Map