Assay Details
| Gene Symbol: Tbx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032005.4 | NP_114394.1 | 2 - 3 | 674 | 99 | |
| GenBank mRNA | AF345520.1 | - | 2 - 3 | 500 | 99 | |
| AF348321.1 | - | 2 - 3 | 631 | 99 | ||
| AK038666.1 | - | 2 - 3 | 676 | 99 | ||
| BC120635.1 | - | 2 - 3 | 545 | 40129558 | 99 | |
| BC137731.1 | - | 2 - 3 | 545 | 8860750 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbx19
Genomic Map