Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 15 - 16 | 1415 | 112 | |
| NM_009549.3 | NP_033575.3 | 15 - 16 | 1418 | 112 | ||
| GenBank mRNA | AK014497.1 | - | 4 - 5 | 466 | 112 | |
| AK048188.1 | - | 15 - 16 | 1396 | 112 | ||
| BC039772.1 | - | 12 - 13 | 1003 | 4191053 | 112 | |
| EF494744.1 | - | 15 - 16 | 1398 | 112 | ||
| U46687.1 | - | 14 - 15 | 1244 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp185
Genomic Map