Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 17 - 18 | 1583 | 76 | |
| NM_009549.3 | NP_033575.3 | 17 - 18 | 1586 | 76 | ||
| GenBank mRNA | AK014497.1 | - | 6 - 7 | 634 | 76 | |
| AK048188.1 | - | 17 - 18 | 1564 | 76 | ||
| BC039772.1 | - | 14 - 15 | 1171 | 4191053 | 76 | |
| EF494744.1 | - | 17 - 18 | 1566 | 76 | ||
| U46687.1 | - | 16 - 17 | 1412 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp185
Genomic Map