Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 1 - 2 | 186 | 122 | |
| NM_009549.3 | NP_033575.3 | 1 - 2 | 186 | 122 | ||
| GenBank mRNA | AK048188.1 | - | 1 - 2 | 166 | 122 | |
| BB648025.1 | - | 1 - 2 | 166 | 122 | ||
| BB663121.1 | - | 1 - 2 | 186 | 122 | ||
| EF494744.1 | - | 1 - 2 | 166 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048188.1, EF494744.1, NM_001109043.1, NM_009549.3 transcripts.
Set Membership: |
|

Zfp185
Genomic Map