Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109043.1 | NP_001102513.1 | 7 - 8 | 653 | 79 | |
| NM_009549.3 | NP_033575.3 | 7 - 8 | 653 | 79 | ||
| GenBank mRNA | BC039772.1 | - | 4 - 5 | 238 | 4191053 | 79 |
| EF494744.1 | - | 7 - 8 | 633 | 79 | ||
| U46687.1 | - | 6 - 7 | 479 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC039772.1, U46687.1 transcripts.
Set Membership: |
|

Zfp185
Genomic Map