Assay Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009549.3 | NP_033575.3 | 9 - 10 | 822 | 92 | |
| GenBank mRNA | BC039772.1 | - | 6 - 7 | 407 | 4191053 | 92 |
| EF494744.1 | - | 9 - 10 | 802 | 92 | ||
| U46687.1 | - | 8 - 9 | 648 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp185
Genomic Map