Assay Details
| Gene Symbol: Vps4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009190.2 | NP_033216.2 | 4 - 5 | 644 | 89 | |
| GenBank mRNA | AK151023.1 | - | 4 - 5 | 598 | 89 | |
| AK152131.1 | - | 4 - 5 | 631 | 89 | ||
| AK165594.1 | - | 589 | 89 | |||
| AK168270.1 | - | 4 - 5 | 535 | 89 | ||
| BC003799.1 | - | 4 - 5 | 551 | 3488471 | 89 | |
| U10119.1 | - | 4 - 5 | 595 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vps4b
Genomic Map