Assay Details
| Gene Symbol: Vps4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009190.2 | NP_033216.2 | 8 - 9 | 1155 | 67 | |
| GenBank mRNA | AK151023.1 | - | 8 - 9 | 1109 | 67 | |
| AK152131.1 | - | 8 - 9 | 1142 | 67 | ||
| AK165594.1 | - | 1100 | 67 | |||
| AK168270.1 | - | 8 - 9 | 1046 | 67 | ||
| BC003799.1 | - | 8 - 9 | 1062 | 3488471 | 67 | |
| U10119.1 | - | 8 - 9 | 1106 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vps4b
Genomic Map