Assay Details
| Gene Symbol: Phlpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133821.3 | NP_598582.3 | 9 - 10 | 2811 | 106 | |
| GenBank mRNA | AB093251.1 | - | 9 - 10 | 1570 | 106 | |
| BC059254.1 | - | 9 - 10 | 1471 | 6406203 | 106 | |
More Information
Set Membership: |
|

Phlpp1
Genomic Map