| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100031599.mm.1 |
NM_001112702.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031599.mm.1 |
NM_001112702.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031599.mm.1 |
NM_021467.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031599.mm.1 |
NM_021467.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100031599.mm.1 |
TC0100003869.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010318.mm.1 |
NM_001112702.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010318.mm.1 |
NM_001112702.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010318.mm.1 |
NM_021467.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010318.mm.1 |
NM_021467.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010318.mm.1 |
TC0100003869.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010321.mm.1 |
NM_001112702.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010321.mm.1 |
NM_001112702.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010321.mm.1 |
NM_021467.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010321.mm.1 |
NM_021467.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010321.mm.1 |
TC0100003869.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010308.mm.1 |
TC0100003869.mm.1 |
MTA TC |