Assay Details
| Gene Symbol: Tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112702.1 | NP_001106173.1 | 7 - 8 | 376 | 58 | |
| NM_021467.5 | NP_067442.1 | 7 - 8 | 424 | 58 | ||
| GenBank mRNA | AA230582.1 | - | 7 - 8 | 424 | 58 | |
| AA271187.1 | - | 7 - 8 | 376 | 58 | ||
| AJ242874.1 | - | 7 - 8 | 424 | 58 | ||
| AK012258.1 | - | 7 - 8 | 407 | 58 | ||
| BC023170.1 | - | 6 - 7 | 488 | 4949804 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnni1
Genomic Map