Assay Details
| Gene Symbol: Ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127346.1 | NP_001120818.1 | 1 - 2 | 181 | 98 | |
| GenBank mRNA | AB183434.1 | - | 1 - 2 | 125 | 98 | |
| AK007894.1 | - | 1 - 2 | 172 | 98 | ||
| AK166492.1 | - | 1 - 2 | 148 | 98 | ||
| AY262012.1 | - | 1 - 2 | 97 | |||
| BC048512.1 | - | 1 - 2 | 140 | 98 | ||
| BC116975.1 | - | 1 - 2 | 147 | 40126294 | 98 | |
| BC116977.1 | - | 1 - 2 | 147 | 40126296 | 98 | |
| BC144716.1 | - | 1 - 2 | 147 | 98 | ||
| BY251841.1 | - | 1 - 2 | 182 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufaf2
Genomic Map