Assay Details
| Gene Symbol: Igfbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008344.3 | NP_032370.2 | 2 - 3 | 832 | 66 | |
| GenBank mRNA | AK157982.1 | - | 2 - 3 | 515 | 66 | |
| BC012723.1 | - | 2 - 3 | 539 | 4224052 | 66 | |
| CT010184.1 | - | 2 - 3 | 470 | 66 | ||
| X81584.1 | - | 2 - 3 | 662 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp6
Genomic Map