Assay Details
| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013566.2 | NP_038594.2 | 3 - 4 | 490 | 102 | |
| GenBank mRNA | AK078871.1 | - | 3 - 4 | 491 | 102 | |
| AK144773.1 | - | 3 - 4 | 462 | 102 | ||
| AK154766.1 | - | 3 - 4 | 491 | 102 | ||
| AK155207.1 | - | 3 - 4 | 457 | 102 | ||
| AK155594.1 | - | 3 - 4 | 462 | 102 | ||
| BC011184.1 | - | 3 - 4 | 463 | 4187781 | 102 | |
| M68903.1 | - | 3 - 4 | 438 | 99 | ||
| M95632.1 | - | 3 - 4 | 434 | 102 | ||
| M95633.1 | - | 3 - 4 | 434 | 102 | ||
| S44607.1 | - | 2 - 3 | 402 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb7
Genomic Map