Assay Details
| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013566.2 | NP_038594.2 | 9 - 10 | 1399 | 64 | |
| GenBank mRNA | AK078871.1 | - | 9 - 10 | 1400 | 64 | |
| AK144773.1 | - | 9 - 10 | 1374 | 64 | ||
| AK154766.1 | - | 9 - 10 | 1400 | 64 | ||
| AK155207.1 | - | 9 - 10 | 1366 | 64 | ||
| AK155594.1 | - | 9 - 10 | 1372 | 64 | ||
| BC011184.1 | - | 9 - 10 | 1372 | 4187781 | 64 | |
| M95632.1 | - | 9 - 10 | 1343 | 64 | ||
| M95633.1 | - | 9 - 10 | 1343 | 64 | ||
| S44607.1 | - | 8 - 9 | 1311 | 64 | ||
More Information
Set Membership: |
|

Itgb7
Genomic Map