Assay Details
| Gene Symbol: Atf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146065.1 | NP_666177.1 | 7 - 8 | 744 | 64 | |
| GenBank mRNA | AK039648.1 | - | 736 | 64 | ||
| AK047901.1 | - | 720 | 64 | |||
| AK154588.1 | - | 735 | 64 | |||
| AK155046.1 | - | 7 - 8 | 735 | 64 | ||
| AK157628.1 | - | 774 | 64 | |||
| BC026483.1 | - | 7 - 8 | 744 | 4503463 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atf7
Genomic Map