Assay Details
| Gene Symbol: Rarg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042727.2 | NP_001036192.1 | 4 - 5 | 890 | 74 | |
| NM_011244.4 | NP_035374.3 | 6 - 7 | 1128 | 74 | ||
| GenBank mRNA | AK148064.1 | - | 6 - 7 | 1125 | 74 | |
| AK155911.1 | - | 4 - 5 | 743 | 74 | ||
| BC012923.1 | - | 6 - 7 | 1054 | 3154037 | 74 | |
| M34475.1 | - | 4 - 5 | 666 | 74 | ||
| M34476.1 | - | 6 - 7 | 885 | 74 | ||
| X15848.1 | - | 6 - 7 | 948 | 74 | ||
More Information
Set Membership: |
|

Rarg
Genomic Map