Assay Details
| Gene Symbol: Rarg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042727.2 | NP_001036192.1 | 5 - 6 | 1066 | 101 | |
| NM_011244.4 | NP_035374.3 | 7 - 8 | 1304 | 101 | ||
| GenBank mRNA | AK148064.1 | - | 7 - 8 | 1301 | 101 | |
| AK155911.1 | - | 5 - 6 | 919 | 101 | ||
| BC012923.1 | - | 7 - 8 | 1230 | 3154037 | 101 | |
| BC013709.1 | - | 5 - 6 | 3989967 | 101 | ||
| M34475.1 | - | 5 - 6 | 842 | 101 | ||
| M34476.1 | - | 7 - 8 | 1061 | 101 | ||
| X15848.1 | - | 7 - 8 | 1124 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rarg
Genomic Map