Assay Details
| Gene Symbol: Csad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144942.4 | NP_659191.1 | 4 - 5 | 509 | 75 | |
| GenBank mRNA | AK005015.1 | - | 4 - 5 | 510 | 75 | |
| AY033912.1 | - | 4 - 5 | 426 | 75 | ||
More Information
Set Membership: |
|

Csad
Genomic Map