Assay Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197940.2 | NP_922922.1 | 4 - 5 | 684 | 72 | |
| GenBank mRNA | AK144539.1 | - | 3 - 4 | 1067 | 72 | |
| BC057854.1 | - | 4 - 5 | 473 | 6494304 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wipf2
Genomic Map