Assay Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197940.2 | NP_922922.1 | 5 - 6 | 1346 | 67 | |
| GenBank mRNA | BC057854.1 | - | 5 - 6 | 1135 | 6494304 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Wipf2
Genomic Map