Assay Details
| Gene Symbol: Ddx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017397.3 | NP_059093.3 | 1 - 2 | 448 | 72 | |
| GenBank mRNA | AF217000.1 | - | 1 - 2 | 323 | 72 | |
| AF217001.1 | - | 1 - 2 | 323 | 72 | ||
| AF220454.1 | - | 1 - 2 | 451 | 72 | ||
| AJ250027.1 | - | 1 - 2 | 323 | 72 | ||
| AK050950.1 | - | 1 - 2 | 322 | 72 | ||
| AK154518.1 | - | 1 - 2 | 327 | 72 | ||
| BC125471.1 | - | 1 - 2 | 448 | 40129986 | 72 | |
| BC137721.1 | - | 1 - 2 | 448 | 8860740 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx20
Genomic Map