Assay Details
| Gene Symbol: Ddx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017397.3 | NP_059093.3 | 3 - 4 | 712 | 109 | |
| GenBank mRNA | AF217000.1 | - | 3 - 4 | 587 | 109 | |
| AF217001.1 | - | 3 - 4 | 587 | 109 | ||
| AF220454.1 | - | 3 - 4 | 715 | 109 | ||
| AJ250027.1 | - | 3 - 4 | 587 | 109 | ||
| AK050950.1 | - | 3 - 4 | 586 | 109 | ||
| AK154518.1 | - | 3 - 4 | 591 | 109 | ||
| BC125471.1 | - | 3 - 4 | 712 | 40129986 | 109 | |
| BC137721.1 | - | 3 - 4 | 712 | 8860740 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx20
Genomic Map