Assay Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027432.3 | NP_081708.1 | 1 - 2 | 201 | 68 | |
| GenBank mRNA | AK012014.1 | - | 1 - 2 | 192 | 68 | |
| AK048488.1 | - | 1 - 2 | 224 | 68 | ||
| AK050391.1 | - | 1 - 2 | 201 | 68 | ||
| AK078856.1 | - | 2 - 3 | 160 | 68 | ||
| AK163839.1 | - | 196 | 68 | |||
| AK168006.1 | - | 1 - 2 | 203 | 68 | ||
| AK168752.1 | - | 1 - 2 | 223 | 68 | ||
| AK169128.1 | - | 1 - 2 | 202 | 68 | ||
| BC005755.1 | - | 1 - 2 | 156 | 2654151 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr77
Genomic Map