Assay Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027432.3 | NP_081708.1 | 3 - 4 | 533 | 84 | |
| GenBank mRNA | AK012014.1 | - | 3 - 4 | 524 | 84 | |
| AK032897.1 | - | 1 - 2 | 72 | 84 | ||
| AK048488.1 | - | 3 - 4 | 556 | 84 | ||
| AK050391.1 | - | 3 - 4 | 533 | 84 | ||
| AK078856.1 | - | 4 - 5 | 492 | 84 | ||
| AK131608.1 | - | 2 - 3 | 444 | 84 | ||
| AK161812.1 | - | 3 - 4 | 508 | 84 | ||
| AK163839.1 | - | 528 | 84 | |||
| AK168006.1 | - | 3 - 4 | 535 | 84 | ||
| AK168752.1 | - | 3 - 4 | 555 | 84 | ||
| AK169128.1 | - | 3 - 4 | 534 | 84 | ||
| BC005755.1 | - | 3 - 4 | 488 | 2654151 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131608.1 transcripts.
Set Membership: |
|

Wdr77
Genomic Map